Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ginsenoside Re

Catalog No. ICN19887905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19887905 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19887905 Supplier MP Biomedicals™ Supplier No. 0219887905
Add to Cart
Add to Cart

From Ginseng root

Reference grade. Purity: ∽95%.

Ref.: Shibata, S., et al., Pharmacol. Bull, 14: 595, (1966).

TRUSTED_SUSTAINABILITY

Specifications

Purity 95%
Content And Storage 2°C to 8°C
Molecular Weight (g/mol) 1079.3
Product Type Ginsenoside Re
Quantity 5 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.