Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ γ-Globulin from Bovine Plasma

Catalog No. ICN19147805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19147805 5 g
ICN19147825 25 g
ICN19147880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19147805 Supplier MP Biomedicals™ Supplier No. 0219147805
Add to Cart
Add to Cart

Purity: 98%

White, crystalline, lyophilized powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 25mg/mL
For Use With (Application) Cell Biology and Analysis, Genomics, Molecular Biology
Formulation The product is lyophilized, in normal Saline solution (25mg/mL, 0.9%NaCl)
Molecular Weight (g/mol) ∽150000
Name g-Globulin from Bovine Plasma
pH Range 6 to 8
Quantity 5 g
Source Bovine Plasma
Storage Requirements 0°C
Form Lyophilized
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.