Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Goat Serum: Normal, Sterile

Catalog No. ICN19135691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Product Type:
Goat Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ICN19135691 1 L Goat Serum
ICN19135680 100 mL Goat Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19135691 Supplier MP Biomedicals™ Supplier No. 0219135691
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable replacement for FBS on most cell lines and primary cultures for Biomedical Research

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance U.S. regulations
Color Red
Content And Storage -20°C
Species Goat
Product Type Goat Serum
Quantity 1 L
Sterility Sterile
Sterilization Method 0.2 μm filtration
For Use With (Application) Immunohistochemistry
Hemoglobin Concentration <20mg/dL
Mycoplasma Detected No
pH 6.5 to 8
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.