Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ground Wheat Bran

Catalog No. ICN10468101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10468101 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10468101 Supplier MP Biomedicals™ Supplier No. 0210468101

Use these wheat middlings for a variety of applications

  • Presentation: Tan flakes
  • CAS: 68916-76-7
TRUSTED_SUSTAINABILITY

Specifications

Product Type Bran
Quantity 1 kg
Storage Requirements Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.