Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Guinea Pig Complement

Catalog No. ICN642831 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN642831 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN642831 Supplier MP Biomedicals™ Supplier No. 08642831
Only null left
Add to Cart
Add to Cart

Specifications

For Use With (Application) Immunoassay
Name Guinea Pig Complement
Quantity 10 mL
Source Guinea Pig
Storage Requirements 2° to 8°C
Cross Reactivity Guinea Pig
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.