Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Gypsy Moth Diet

Catalog No. ICN96029301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
4 L
100 L
Format:
Agar packaged separately
Premixed with agar
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
ICN96029301 1 L Agar packaged separately
ICN96029201 1 L Premixed with agar
MP296029304 4 L Agar packaged separately
MP296029204 4 L Premixed with agar
MP296029380 100 L Agar packaged separately
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN96029301 Supplier MP Biomedicals™ Supplier No. 0296029301
Add to Cart
Add to Cart

Used for general colony maintenance of moth larvae

Applications:
Gypsy Moth Diet is used for general colony maintenance of moth larvae

Product Description:
Gypsy moth diet has previously been known as the High Wheat Germ Diet. The Gypsy Moth Diet is available with agar included, packaged separately or omitted completely. Methyl paraben and sorbic acid added as preservatives. For complete diet composition, please see the Product Datasheet

Recommended Use:
Add 60 grams of Agar to approximately 3.6 liters of distilled water. Bring to a boil. Allow to dissolve. Add 664 grams of Gypsy Moth Diet to the boiling Agar solution. Add water if necessary to bring to volume. Add antimicrobial agents at this point if desired. Mix the formulation well in a large blender before dispensing into rearing containers.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Agar packaged separately
For Use With (Application) Gypsy Moth diet is used for general colony maintenance of moth larvae
Quantity 1 L
Storage Requirements 15°C to 30°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.