Learn More
MP Biomedicals™ 1X Hanks' Balanced Salt Solution (Modified) without Magnesium and Calcium
Shop All MP Biomedicals, Inc ProductsDescription
Specifications
Specifications
| Chemical Name or Material | Hanks' Balanced Salt Solution (HBSS) |
| Color | Red |
| Concentration | 1 X |
| pH | 6.9 to 7.4 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Formulation | Potassium Chloride 400 mg/L, Potassium Phosphate Monobasic 60 mg/L, Sodium Bicarbonate 350 mg/L, Sodium Chloride 8000 mg/L, Sodium Phosphate Dibasic 47.50 mg/L, Dextrose 1000 mg/L, Phenol Red.Sodium Salt 17 mg/L |
| Grade | Not Available |
Safety and Handling
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.