Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hat Medium

Catalog No. ICN1680849 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1680849 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1680849 Supplier MP Biomedicals™ Supplier No. 091680849
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the selective growth of hybridoma cells for monoclonal antibody production.

TRUSTED_SUSTAINABILITY

Specifications

Product Type HAT medium Supplement
Sterility Sterile
Quantity 100 mL
Content And Storage 0° to -20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.