Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hemoglobin from Bovine Erythrocytes

Catalog No. p-4605361 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10071425 25 g
ICN10071491 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10071425 Supplier MP Biomedicals™ Supplier No. 0210071425
Only null left
Add to Cart
Add to Cart

Acts as oxygen transporter

Hemoglobin is the major component of red blood cells, and is responsible for their red color. A major oxygen-transporting component of red blood cells Its normal concentration in erythrocytes is 34%. Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide. Hemoglobin is a tetramer composed of 2 pairs of polypeptide chains called globins, and 4 heme groups. Each polypeptide chain is bound to one heme. Iron is coordinated to 4 pyrrole nitrogens of protoporphyrin IX and to an imidazole nitrogen of a histidine residue from the globin side of the porphyrin.

  • CAS Number 9008-02-0
  • MDL Number MFCD00131282
  • Format: Reddish-brown crystalline powder
  • Moisture content: <10%
  • pH: 7.0 to 9.0 (2% solution)
  • Soluble in water (1 part in 7 of water) and slowly soluble in glycerol

Used to study paradigm for regulation of gene expression and cellular differentiation.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Biology and Analysis, Cell Culture, Genomics, Molecular Biology
Molecular Weight (g/mol) ∽64.5
Name Hemoglobin
pH Range 7 to 9
Quantity 25 g
Source Bovine
Storage Requirements 4°C
Form Crystalline
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.