Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hormones: Dexamethasone

Catalog No. ICN19456191 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1000 mg
25 mg
500 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19456191 1000 mg
ICN19456125 25 mg
ICN19456180 100 mg
ICN19456190 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19456191 Supplier MP Biomedicals™ Supplier No. 0219456191
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to study apoptosis, cell signaling pathways and gene expression. MP Biomedicals Hormones: Dexamethasone acts as anti-inflammatory glucocorticoid that induces the production of phospholipase A2 inhibitory protein (lipocortin).

  • Inhibits induction of nitric oxide synthase (IC50=5 nM)
  • Causes reduction in cyclin A and Cdk2 activity, inhibition of G1/S transition in osteoblasts and inhibition of phosphorylation of Rb protein
  • Observed to induce apoptosis in human thymocytes and eosinophils but to inhibit apoptosis in neutrophils
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Form Crystalline powder
Product Type Dexamethasone
Quantity 1000 mg
WARNING: Cancer - www.P65Warnings.ca.gov

For research use only. Not intended for therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.