Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hormones: Insulin

Catalog No. ICN10570725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10570725 25 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10570725 Supplier MP Biomedicals™ Supplier No. 10570725

missing translation for 'validMainframeMsg'

For research use only. Not intended for therapeutic use.

†Zinc stabilized.

‡Zinc content: 0.25 to 1.1%.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Insulin
Quantity 25 kU
Source Bovine pancreas
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.