Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Human Albumin, 30%

Catalog No. ICN823051 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN823051 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN823051 Supplier MP Biomedicals™ Supplier No. 08823051
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

30% Solution prepared from Fraction V human albumin

  • CAS Number: 70024-90-7
  • EC Number: 274-272-6
  • MDL Number: MFCD00081418
  • Format: solution
  • Isoelectric point: 4.7
  • Supplied at pH 7, without any preservative
  • Sterile with purity >96%

For immunoassays and mammalian cell culture

TRUSTED_SUSTAINABILITY

Specifications

Concentration 30g/dL
For Use With (Application) Immunoassay, Mammalian Cell Culture
Formulation Supplied at a pH 7.1 ±0.3 and contains 0.1% Sodium Azide as a preservative
Name Human Albumin (30% Solution)
pH Range 7.1
Quantity 50 mL
Source Human
Storage Requirements 2° to 8°C
Form Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.