Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Interleukin 2, Human, Recombinant

Catalog No. ICN15389610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kU/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15389610 10 kU/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15389610 Supplier MP Biomedicals™ Supplier No. 0215389610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Interleukin 2, Human, Recombinant plays a role in the activation and proliferation of NK cells, induces γ-interferon and B cell growth factor secretion, and modulates the expression of the IL-2 receptor.

  • Interleukin-2 plays a role in the activation and proliferation of NK cells, induces γ-inter­feron and B cell growth factor secretion, and modulates the expression of the IL-2 receptor.
TRUSTED_SUSTAINABILITY

Specifications

Endotoxin Level <0.1 ng/mg
For Use With (Application) For supplementing cell cultures and investigating various metabolic processes
Purity >98% by SDS-PAGE analysis
Quantity 10 kU/L
Sterility Sterile
Content And Storage -20°C
Reactivity Human

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.