Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Human Serum: Heat Activated

Catalog No. ICN2930249 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN2930249 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN2930249 Supplier MP Biomedicals™ Supplier No. 092930249

missing translation for 'validMainframeMsg'

Hepatitis Associated Antigen (HBsAG)–free, HIV–free

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance U.S. regulations
Species Human
Product Type Human Serum
Quantity 100 mL
Heat Inactivated Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.