Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Serum, Type AB, Human (HCV, HIV, HbsAG Free)

Catalog No. ICN2931949 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN2931949 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN2931949 Supplier MP Biomedicals™ Supplier No. 092931949
Only null left
Add to Cart
Add to Cart

Used as a growth supplement for cell culture media and in other research applications.

  • Collected from type AB human blood
  • Used as a growth supplement for cell culture media and in other research applications
  • Proven to grow many human cell lines at a faster rate than mixed blood group serum

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance U.S. regulations
Color Golden Brown
Content And Storage -20°C
Species Human
Form Liquid
Product Type Human Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method 0.1 μm filtration
For Use With (Application) Grow many Human cell lines at a faster rate
Hemoglobin Concentration ≤26 mg/dL
Mycoplasma Detected No
Osmolality 263 to 323 mOsm/kg
pH 7 to 8
Protein Content 5.5 to 7.5 g/dL
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.