Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Immuno™ Albumin, Bovine, Low Endotoxin Powder (Heat-Shock)

Catalog No. MP08810681 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08810681 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08810681 Supplier MP Biomedicals™ Supplier No. 08810681
Add to Cart
Add to Cart

Used to solubilize lipids, also used as blocking agent in Western blots or ELISA applications.

  • Derived from bovine blood collected at USDA licensed facility
  • Prepared by heat-shocked fractionate method
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Immunoassay, Mammalian Cell Culture, Western Blot
Molecular Weight (g/mol) ∽66
Name Immuno™ Albumin, Bovine, Low Endotoxin (Heat Shock)
pH Range 7.1
Quantity 25 g
Source Bovine Blood
Storage Requirements 0° to 5°C
Form Powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.