Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Joklik's Modification of Minimum Essential Medium Eagle, Liquid

Catalog No. MP091032322 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091032322 1 x 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091032322 Supplier MP Biomedicals™ Supplier No. 091032322

missing translation for 'validMainframeMsg'

For suspension cultures with penicillin, streptomycin, L-glutamine and sodium

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 0° to 5°C
Product Type Minimum Essential Medium Eagle's
Sterility Sterile
pH 6.3 to 7.2
Quantity 1 x 10 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.