Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ L-Broth, Capsules

Catalog No. MP113001031 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
454 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113001031 1 kg
MP113001021 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP113001031 Supplier MP Biomedicals™ Supplier No. 113001031
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the growth of E.coli and certified for plasmid and other bacterial cultures in liquid medium

  • Nutritionally rich medium originally developed for growth and maintenance of E. coli strains
  • Eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder
  • Contains essential electrolytes for transport and osmotic balance, due to NaCl component
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Format Capsules
Grade Molecular Biology Certified™
Media Type Bacterial Media
Product Type Microbiology Media
Sterility Sterile
Formulation 10g Tryptone, 5g Yeast Extract, 0.5g NaCl. Small quantities of biological buffers have been added for proper pH adjustment.
pH 6.9
Quantity 1 kg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.