Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ LB Agar Medium Plates

Catalog No. MP113002234 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
15 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113002234 15 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113002234 Supplier MP Biomedicals™ Supplier No. 113002234
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rich medium for the growth of E. coli. and certified for plasmid and other bacterial cultures in liquid medium

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Manufacturing Origin United States
Media Type LB Agar medium
Formulation Tryptone 10g/L, Yeast 5g/L
For Use With (Application) For growth of Escherichia coli, plasmid and other bacterial cultures
Packaging Type Plate
Quantity 15 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.