Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ LB Media, Sterile Solutions

Catalog No. MP113002014 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP113002014 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113002014 Supplier MP Biomedicals™ Supplier No. 113002014
Add to Cart
Add to Cart

Formulations for the E. coli growth media known as LB Medium typically contain 10g of tryptone and 5g of yeast extract per liter; this formulation also contains 10g NaCl per liter

LB medium is a nutritionally rich, sterile solution, Molecular Biology Certified™ for the growth of E. coli and other bacterial cultures.

  • Supports luxurious growth of E. coli cells.
  • Supplies essential growth factors (tryptone and yeast extract).
  • Contains essential electrolytes (NaCl) for transport and osmotic balance.
  • pH 7.0.
  • Clear, light yellow liquid appearance.
TRUSTED_SUSTAINABILITY

Specifications

Appearance Clear, Light Yellow
Broth Type LB Medium
Certifications/Compliance Molecular Biology Certified
Color Light Yellow
Concentration 25 g/L
Content And Storage 15° to 30°C
Country of Origin United States
Additive Biological Buffers
Detects Bacterial Growth, Plasmid Production
Form Solution
Manufacturing Origin United States
Media Type LB Medium
Product Type Bacterial Growth Medium
Sterility Sterile
Target Organism Class Bacteria
Test Type Growth Promotion Testing
Fill Volume 500 mL
Final Product Type Bacterial Culture, Plasmid
Formulation Contents/L-10 g Tryptone, 5 g Yeast Extract, 10 g NaCl., Biological Buffers
For Use With (Application) Cell Culture, Proteomics, Purification and Detection
pH 7
Quantity 500 mL
Specimen Type E. coli
Protein Source Casein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.