Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Linbro™ 96-Well, Cell Culture-Treated, V-Shaped-Bottom Microplate

Catalog No. ICN7602305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
ICN7602305 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN7602305 Supplier MP Biomedicals™ Supplier No. 097602305

missing translation for 'validMainframeMsg'

Color:Clear; Sterility:Sterile; Lid:With Lid

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Conical Bottom (V-well)
Width (Metric) 8.6 cm
Height (Metric) 1.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.