Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Linbro™ 96-Well Microplates

Catalog No. ICN7622305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
ICN7622305 Conical Bottom (V-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN7622305 Supplier MP Biomedicals™ Supplier No. 097622305

missing translation for 'validMainframeMsg'

Specifications

Format Standard
Lid With Lid
Array 8 x 12
Color Clear
Product Type 96-well Assay Microplate
Material Polystyrene
Volume (Metric) Well 0.25 mL
No. of Wells 96
Sterility Sterile
Surface Treatment Untreated
Well Shape Conical Bottom (V-well)
Width (Metric) 8.6 cm
Height (Metric) 1.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.