Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lipoprotein-Cholesterol Concentrate

Catalog No. ICN19147690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19147690 500 mL
ICN19147625 25 mL
ICN19147680 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19147690 Supplier MP Biomedicals™ Supplier No. 0219147690

From bovine serum

A mixture of lipoprotein cholesterol, phospholipids and fatty acids. For use as a serum-free or serum-reduced media nutrient supplement, calibration of cholesterol values, and enzymatic or nonenzymatic cholesterol determinations. Offered as a low-salt solution. No preservatives. Free of enzyme inhibitors. Filtered through 0.2μm membrane filter. Cholesterol concentration: 1000-2000mg/dL.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1000 to 2000mg/dL
For Use With (Application) Media Nutrient Supplement, Calibration of Cholesterol Values, Enzymatic or Non-enzymatic Cholesterol Determinations
Name Lipoprotein Cholesterol concentrate
pH Range 7.5 to 8
Quantity 500 mL
Source Bovine Serum
Storage Requirements 0° to 5°C
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.