Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lysing Matrix D, Bulk
GSA_VA

Catalog No. 116540434 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-654-0434 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-654-0434 Supplier MP Biomedicals™ Supplier No. 116540434
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lysing Matrix D Bulk contains 1.4 mm ceramic spheres and are found in the FastDNA™ Green Spin Kit for isolation of genomic DNA from plant and animal tissue and the FastRNA™ Pro Green Kit for isolation of total RNA from plant and animal tissues.

  • Product Overview: Lysing Matrix D bulk contains 1.4 mm ceramic spheres and are found in the FastDNA™ Green Spin Kit for isolation of genomic DNA from plant and animal tissue and the FastRNA™ Pro Green Kit for isolation of total RNA from plant and animal tissues.
  • Applications: Lysing Matrix D is used primarily for lysis of softer tissues like brain, liver, kidney, lung, and spleen. It can also used to lyse plant tissue such a leaves, roots, and fruits, as well as cultured cells and whole insects. This grinding matrix is chemically inert and will not bind nucleic acids.
  • Sample Type: Animal Tissues, Plants, Cultured Cells

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Isolation Technology Mechanical Disruption
Sample Type Animal Tissues, Plants, Cultured Cells, Insects, Worms
For Use With (Equipment) FastPrep-24; FastPrep 120; FastPrep-96
For Use With (Application) Nucleic Acid Isolation and Purification
Matrix Lysing Matrix D
Product Type Lysing matrix
Quantity 500 g

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.