Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ TeenPrep™ Lysing Matrix F, 15 mL Tubes

Catalog No. MP116935025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 x 15 mL Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116935025 25 x 15 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116935025 Supplier MP Biomedicals™ Supplier No. 116935025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TeenPrep™ Lysing Matrix F, 15 mL Tubes are used to isolate RNA and protein from samples. Lysing Matrix F is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.

  • Impact-resistant 15 mL tubes contain 1.6 mm aluminum oxide particles and 1.6 mm silicon carbide particles
  • With white caps
  • Specific Gravity: Aluminum Oxide, 3.95; Silicon Carbide, 3.20
  • Nucleic Acid Isolation and Purification
TRUSTED_SUSTAINABILITY

Specifications

Capacity 15 mL
Format Tube
Isolation Technology Mechanical Disruption
Sample Type Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold
DNase Dnase-free
For Use With (Application) Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Matrix Lysing matrix F
Molecular Formula Aluminum Oxide, Al2O3; Silicon Carbide, SiC
Product Type Lysing Matrix
Quantity 25 x 15 mL Tubes
RNase RNase-free
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.