Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lysing Matrix G

Catalog No. MP116916100 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 x 2 mL Tubes
50 x 2 mL Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116916100 100 x 2 mL Tubes
MP116916050 50 x 2 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP116916100 Supplier MP Biomedicals™ Supplier No. 116916100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lysing Matrix G Tubes for optimal lysis used with FastPrep™ instruments.

Silicon carbide has relatively low density and high strength and hardness, and therefore is effective at breaking tough, hard, or brittle cell membranes. Useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.

  • Impact-resistant 2 mL tube contains 1.6 mm Silicon Carbide particles and 2 mm glass beads
  • With brown caps
  • Specific Gravity: Silicon Carbide: 3.20; Glass beads: 2.55
  • Nucleic Acid Isolation and Purification
TRUSTED_SUSTAINABILITY

Specifications

Capacity 2 mL
Sample Type Spores, Animal Tissues, Fungi, Plants, Yeast
DNase Dnase-free
For Use With (Application) Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Matrix Lysing matrix G
Molecular Formula Silicon Carbide, SiC ; Glass, SiO2
Quantity 100 x 2 mL Tubes
RNase RNase-free
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.