Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ M9 Minimal Medium Salts (Maniatis Formulation), Powdered

Catalog No. MP113037012 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
227 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113037012 227 g
MP113037032 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP113037012 Supplier MP Biomedicals™ Supplier No. 113037012
Add to Cart
Add to Cart

Used for preparing a defined synthetic medium for the growth of E. coli host

  • A commonly used base for preparing a defined synthetic medium for the growth of most E. coli host cells
  • Powder is reconstituted in sterile purified water, (or water with melted purified agar added)
  • Additional nutrients, such as glucose or other carbon source, casamino acids, vitamins, or antibiotics are added when the solution cools
  • Add additional nutrients before plates are poured or media is inoculated with bacteria
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified
Concentration 56.4 g/L of a 5X Solution
Content And Storage 15° to 30°C
Description A 56.4 g sample of M9 Minimal Salts was dissolved in an aqueous solution to 1 liter, the pH adjusted to 7.4 (if needed), and the solution autoclaved to make a sterile 5X concentrate.
Form Powder
Media Type M9 Minimal Medium
Product Type Microbiology Media
Sterility Sterile
Formulation 33.9 g Na2HPO4; 15 g KH2PO4; 2.5 g NaCl; 5 g NH4Cl
For Use With (Application) Preparing a Defined Synthetic Medium for The Growth of E. Coli Host
pH 6.8
Quantity 227 g
Shipping Condition Ambient
Solubility Soluble Aqueous Solution
Specimen Type Bacterial
Strain Designations E. coli
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.