Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Menhaden Oil

Catalog No. ICN96012001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN96012001 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN96012001 Supplier MP Biomedicals™ Supplier No. 0296012001
Add to Cart
Add to Cart

Menhaden Oil, refined

Menhaden oil is use as a control or to check sample for fatty acid methyl ester (FAME) analyses. It is rich in omega-3 fatty acids. When used to partially supplement a high-fat diet in diabetic rats, Menhaden oil improved end points for peripheral neuropathy by improving motor and sensory nerve conduction velocity. Decreased accumulation of fatty acids in rat livers was also demonstrated.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Lyophilized
For Use With (Application) Menhaden Oil is use as a control or check sample for Fatty Acid Methyl Ester (FAME) analyses
Quantity 1 gal.
Storage Requirements -20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.