Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Merrifield's Peptide Resin

Catalog No. ICN15160880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15160880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15160880 Supplier MP Biomedicals™ Supplier No. 0215160880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sieve Size of 200 to 400 mesh or 40 to 80μm

TRUSTED_SUSTAINABILITY

Specifications

Color White
Format Powder
Product Type Merrifields Peptide Resin
Quantity 100 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.