Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Miniprep Express™ Matrix

Catalog No. 112000200 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
1250 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
11-200-0200 1250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-200-0200 Supplier MP Biomedicals™ Supplier No. 112000200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for rapid purification of plasmid DNA. Use Miniprep Express™ Matrix to quickly screen hundreds of recombinant clones in record time using restriction enzyme analysis and PCR.

  • MiniPrep Express™ Matrix features an ultra-streamlined protocol resulting in high-quality plasmid DNA
  • Provides rapid purification and allows screening of up to 1250 samples for pennies per prep
  • MiniPrep Express™ Matrix provides the essential ingredient for making your ''own kit'' for doing large numbers of minipreps for rapid screening purposes that involve a restriction digest
  • This kit allows you to make the lysis reagents from the most commonly used methods
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Format Kit
Isolation Technology Alkaline Lysis
Sample Type Plasmid Prep
For Use With (Application) Rapid purification of plasmid DNA, Nucleic Acid Isolation and Purification
Includes MiniPrep Express™ Matrix, Protocols and recipes necessary for either alkaline lysis or boiling miniprep procedures.
No. of Reactions 1250 Preps
Product Type Plasmid Purification Kit
Sample Volume 1 mL of Overnight Culture
Synonym Plasmid Miniprep Purification Kit
Yield Yields 1 to 5 μg of High Copy Number Vector DNA Per 1 mL overnight culture typical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.