Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Mouse Serum

Catalog No. ICN15228201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
Form:
Liquid
Lyophilized
Hemoglobin Concentration:
5.8 g/dL
9.65 mg/dL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Hemoglobin Concentration
ICN15228201 1 mL Liquid 9.65 mg/dL
ICN15228205 5 mL Liquid 9.65 mg/dL
ICN642931 10 mL Lyophilized 5.8 g/dL
ICN15228105 - - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15228201 Supplier MP Biomedicals™ Supplier No. 0215228201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for blocking agents and controls in immunoassays.

TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Content And Storage -20°C
Species Mouse
Form Liquid
Product Type Mouse Serum
Quantity 1 mL
Sterilization Method 0.2 μm filtration
For Use With (Application) Blocking agents and controls in immunoassays
Hemoglobin Concentration 9.65 mg/dL
pH 7.4 to 8.1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.