Learn More
Specifications
Specifications
| Color | Brown |
| Content And Storage | -20°C |
| Species | Mouse |
| Form | Liquid |
| Product Type | Mouse Serum |
| Quantity | 5 mL |
| Sterilization Method | 0.2 μm filtration |
| For Use With (Application) | Blocking agents and controls in immunoassays |
| Hemoglobin Concentration | 9.65 mg/dL |
| pH | 7.4 to 8.1 |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.