Learn More
Description
- MT Buffer is a lysis solution component included in the FastRNAPro Soil Direct and Indirect Kits, the FastDNA Spin Kit for Feces, and the FastDNA 50 mL Spin Kit for Soil.
- Used as a stabilization agent during sample lysis.
- Homogenization takes place in the presence of MT Buffer, this reagent is carefully developed to protect and solubilize nucleic acids and proteins upon cell lysis.
- Thus allowing extraction of genomic DNA with minimal RNA contamination.
Specifications
Specifications
| Format | Bottle |
| Sample Type | Feces, Soil, Wastewater, Environmental Water, Sludge |
| For Use With (Equipment) | FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1 |
| For Use With (Application) | Sample preparation, sample lysis, nucleic acid isolation and purification, molecular biology, gene expression, microbiological analysis, gene function, gene regulation |
| Product Type | MT Buffer |
| Quantity | 8 mL |
| Sample Volume | Up to 500 mg Per Prep |
| Sample Source | Microorganisms |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.