Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Sodium Iodide (NaI) Solution

Catalog No. p-4943378 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 60 mL
1 x 125 mL
2 x 125 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP111001201 1 x 60 mL
MP111001401 1 x 125 mL
MP111001601 2 x 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP111001201 Supplier MP Biomedicals™ Supplier No. 111001201
Only null left
Add to Cart
Add to Cart

Sodium Iodide (NaI) Solution dissolves agarose gels and helps in binding the DNA to the Glassmilk™ binding matrix.

  • Less than 15% DNA 1 kb and above is left unbound in NaI.
  • NaI (Sodium Iodide) is supplied with GeneClean™ kits and helps in binding the DNA to the GLASSMILK binding matrix.
  • Melts 0.1 g gel slice in 5 minutes at 55°C.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7681-82-5
Concentration 6 M
Content And Storage 15°C to 30°C
Formula Weight 149.89
For Use With (Application) Nucleic Acid Isolation and Purification
Molecular Formula NaI
Melting Point 661°C
pH 7.0 to 7.4
Product Type NaI
Quantity 1 x 60 mL
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.