Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Wash Concentrate, MP Biomedicals™

Catalog No. MP111001402 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
14 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP111001402 14 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP111001402 Supplier MP Biomedicals, Inc Supplier No. 111001402
Add to Cart
Add to Cart

New Wash Concentrate

  • Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2-8°C
For Use With (Application) Nucleic Acid Isolation & Purification
pH 7.62
Product Type New Wash Concentrate
Quantity 14 mL
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.