Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Nigrosin Stain

Catalog No. ICN19459380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19459380 100 g
ICN19459325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19459380 Supplier MP Biomedicals™ Supplier No. 0219459380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a viable cell staining agent especially in stem cell biology

  • Formula Weight: 737.87
  • CAS # 8005-03-6
  • Synonym: Acid Black 2; C.I. 50420
  • Physical Description: Black crystalline powder
  • Solublility: Soluble in water(1mg/ml-opaque black with purple cast solution)
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Format Crystal
For Use With (Application) Viable cell staining agent especially in stem cell biology
Storage Requirements 15 to 30°C
Product Type Nigrosin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.