Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Nisin

Catalog No. ICN15583925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15583925 25 g
ICN15583905 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15583925 Supplier MP Biomedicals™ Supplier No. 0215583925
Add to Cart
Add to Cart

Nisin content: 2.5% Remainder is NaCl and milk solids (denatured)

  • Molecular Formula: C14H230N42O37S7
  • Molecular Weight: 3354.3

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
pH 3.31
Quantity 25 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.