Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Other Growth Factors and Supplements

Catalog No. ICN19113080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.1 mg
10 μ g
25 mg
Product Type:
B-nerve Growth Factor
Endothelial Cell Growth
Fibroblast Growth Factor
Nerve Growth Factor
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ICN19113080 0.1 mg Nerve Growth Factor
ICN15233025 25 mg Endothelial Cell Growth
ICN15984010 10 μ g B-nerve Growth Factor
ICN16003701 10 μ g Fibroblast Growth Factor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19113080 Supplier MP Biomedicals™ Supplier No. 19113080

missing translation for 'validMainframeMsg'

Suitable for supplementing cell cultures and investigating various metabolic processes

†With streptomycin sulfate.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Nerve Growth Factor
For Use With (Application) Supplementing cell cultures and investigating various metabolic processes
Quantity 0.1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.