Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G Potassium Salt, MP Biomedicals™

Catalog No. ICN19453680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mU
10 mU
100 μm
25 mU
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19453680 100 μm
MP219453601 1 mU
MP219453610 10 mU
MP219453625 25 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19453680 Supplier MP Biomedicals, Inc Supplier No. 0219453680
Add to Cart
Add to Cart

Penicillin G Potassium Salt Cell culture reagent, White Powder

  • It is used as a cell culture additive as an antibiotics.
  • Penicillin G potassium Salt is a salt of Penicillin G.
  • Penicillin G is a hydrophobic, β-lactam antibiotic produced by Penicillium spp.
  • Penicillin G is also known as benzylpenicillin and contains a 6’ phenylacetyl side chain.
  • In acid aqueous solutions, Penicillin is reported to decompose into penillic acid and penilloic acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00036193
Synonym Benzylpenicillin Potassium Salt

Specifications

Physical Form Powder
pH 5 to 7.5 (3% aq. soln.)
Melting Point 214°C to 217°C (Literature)
CAS 113-98-4
Potency 1500 to 1700 μ/mg
Sterility Non-sterile
Color White
Quantity 100 μm
Validated Application Cell Culture
Molecular Formula C16H17N2O4SK
Beilstein 3832841EC
Solubility Information Soluble in water(100mg/mL).
Molecular Weight (g/mol) 372.48
Grade Cell Culture
Assay 0.998
Show More Show Less
RTECSNumber XH9700000
storageNote1 Store at +4°C.
Recommended Storage 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.