Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G Potassium Salt, MP Biomedicals™

Catalog No. MP219453601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mU
10 mU
100 μm
25 mU
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219453601 1 mU
ICN19453680 100 μm
MP219453610 10 mU
MP219453625 25 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219453601 Supplier MP Biomedicals, Inc Supplier No. 0219453601
Only null left
Add to Cart
Add to Cart

Penicillin G Potassium Salt Cell culture reagent, White Powder

  • It is used as a cell culture additive as an antibiotics.
  • Penicillin G potassium Salt is a salt of Penicillin G.
  • Penicillin G is a hydrophobic, β-lactam antibiotic produced by Penicillium spp.
  • Penicillin G is also known as benzylpenicillin and contains a 6’ phenylacetyl side chain.
  • In acid aqueous solutions, Penicillin is reported to decompose into penillic acid and penilloic acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-98-4
MDL Number MFCD00036193
InChI Key HTQQVOYPSGNVPA-HBRCYENSSA-M
Synonym Benzylpenicillin Potassium Salt
IUPAC Name potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Physical Form Powder
pH 5 to 7.5 (3% aq soln)
Melting Point 214°C to 217°C (literature)
Potency 1,500 - 1,700 u/mg
Color White
Quantity 1 mU
Validated Application Cell Culture
Molecular Formula C33H36ClKN4O4S2
Beilstein 3832841EC
Solubility Information Soluble in water(100mg/mL).
Molecular Weight (g/mol) 691.34
Grade Cell Culture
Assay 0.998
Show More Show Less
RTECSNumber XH9700000
storageNote1 Store at +4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.