Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pepsin

Catalog No. ICN19536710 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19536710 10 g
ICN19536705 5 g
ICN19536701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19536710 Supplier MP Biomedicals™ Supplier No. 0219536710
Only null left
Add to Cart
Add to Cart

From porcine stomach mucosa

Purified by chromatography. Activity: 2000-2400U/mg protein. Unit Definition: One unit will produce a Δ A280 of 0.001 per minute at pH 2.0 and 37°C, measured as TCA-soluble products using hemoglobin as substrate. Lyophilized powder, essentially salt-free.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Used to digest Proteins and is commonly used in the preparation of Fab fragments from Antibodies
pH Range 1 to 3.5
Sample Type Porcine Stomach Mucosa
Content And Storage 0°C
Form Lyophilized
Quantity 10 g
Product Type Pepsin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.