Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ PhoenIX™ Gigaprep Kit

Catalog No. MP112075400 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
5 Preparations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP112075400 5 Preparations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112075400 Supplier MP Biomedicals™ Supplier No. 112075400

Designed for fast purification of up to 10mg transfection-grade plasmid or cosmid DNA.

  • Lysate filtration cartridge included for maximum efficiency
  • Easy-to-use
  • patented gravity flow resin in pre-packed column with filter
  • no hazardous buffers or organic reagents
TRUSTED_SUSTAINABILITY

Specifications

No. of Reactions 5 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.