Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ PhoenIX ™ Midiprep Kit

Catalog No. MP112075200 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
25 Preparations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP112075200 25 Preparations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112075200 Supplier MP Biomedicals™ Supplier No. 112075200

Designed for fast purification of up to 100μg transfection-grade plasmid or cosmid DNA.

  • Easy-to-use
  • patented gravity flow resin in pre-packed column with filter
  • no hazardous buffers or organic reagents
TRUSTED_SUSTAINABILITY

Specifications

No. of Reactions 25 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.