Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pipet Tips for Finnpipettes™

Catalog No. ICN7774703 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Volume: 25 to 100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN7774703 Volume: 25 to 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN7774703 Supplier MP Biomedicals™ Supplier No. 097774703

missing translation for 'validMainframeMsg'

Steptip #2, Sterile, 25 to 100 μL Pipette Tips for Finnpipettes

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 25 to 100 L
Sterility Sterile
For Use With (Equipment) Thermo Scientific Finnpipette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.