Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cover for PlantCon™ Plant Tissue Culture Container

Catalog No. ICN2672102 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10/Cs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN2672102 10/Cs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN2672102 Supplier MP Biomedicals™ Supplier No. 092672102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Plastic cover for PlantCon™ Plant Tissue Culture Container. Use PlantCon™ containers with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.

  • Cover can be snapped on tight, for long-term culture, or loosely fitted during acclimation to external conditions prior to transplantation
  • Ideal for use in commercial horticulture or research settings
  • Allow exceptional light transmission and optimal growth
  • Reduce the time spent preparing an in vitro growth environment for plant cultures by eliminating the need for cleaning and sterilization
TRUSTED_SUSTAINABILITY

Specifications

Closure Material Plastic
Bottom Shape Square
Color Clear
Disposable Single-use
Material PVC
Shape Rectangular
Sterility Sterile
Type Cell Culture Dish Cover
For Use With (Application) Optimal Plant Cell Culture Applications
For Use With (Equipment) PlantCon™ Containers (092672206, 092672002)
Quantity 10/Cs
Vent Yes
Dimensions (L x W) 3.75 x 3.75 in. (9.5 x 9.5 cm)
Dimensions (L x W x H) 9.5 x 9.5 x 7 cm
Length (Metric) 9.5 cm
Width (Metric) 9.5 cm
Height (Metric) 7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.