Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Plasminogen from Human Plasma

Catalog No. ICN19134225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
25U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19134225 25U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19134225 Supplier MP Biomedicals™ Supplier No. 0219134225
Only null left
Add to Cart
Add to Cart

Purity: >98% by SDS-PAGE

Unit Definition: One unit is defined as the amount of enzyme which will hydrolyze 1μmol of N-Tosyl-L-Arginine Ester in 30 minutes at 37°C, pH 8.0. Supplied as a lyophilized powder from 0.5mL of 20mM Tris-HCl buffer, 2mM EDTA, pH 7.5.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a lyophilized powder from 0.5mL of 20mM Tris HCl Buffer, 2mM EDTA
Molecular Weight (g/mol) 90000 to 94000
Name Plasminogen
pH Range 7.5
Purification Method SDS-PAGE
Quantity 25U
Source Human Plasma
Storage Requirements 0°C
Form Lyophilized
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.