Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pokeweed Mitogen, Lyoph.

Catalog No. MP08780332 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08780332 1mg
ICN780331 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP08780332 Supplier MP Biomedicals™ Supplier No. 08780332
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phytolacca Americana

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunoassay
Name Pokeweed Mitogen
pH Range 7.2 to 7.4
Quantity 1mg
Storage Requirements 2° to 8°C
Recombinant Recombinant
Form Lyophilized
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.