Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Poly-D-Lysine Hydrobromide Attachment Factors

Catalog No. ICN15017510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15017510 10 mg
ICN15017525 25 mg
ICN15017550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15017510 Supplier MP Biomedicals™ Supplier No. 0215017510
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Facilitate attachment, adherence, and spreading in the culture of adherent cells

Nonsterile.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Poly-D-Lysine
Sterility Non-sterile
Quantity 10 mg
Source Synthetic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.