Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Poly-L-Lysine Hydrobromide Attachment Factors: ≥70,000 MW

Catalog No. ICN15017710 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Sterility:
Non-sterile
Sterile
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sterility
ICN15017710 10 mg Non-sterile
ICN15017780 100 mg Non-sterile
ICN15017725 25 mg Non-sterile
ICN15017750 50 mg Non-sterile
ICN19454405 5 mg Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN15017710 Supplier MP Biomedicals™ Supplier No. 0215017710
Add to Cart
Add to Cart

Facilitate attachment, adherence, and spreading in the culture of adherent cells

Available nonsterile or gamma-irradiated.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Poly-L-Lysine
Sterility Non-sterile
Quantity 10 mg
Source Synthetic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.