Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Protease, from S. aureus strain V8

Catalog No. ICN15197205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15197205 5 mg
ICN15197201 1 mg
ICN15197210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15197205 Supplier MP Biomedicals™ Supplier No. 0215197205
Add to Cart
Add to Cart

Used for selective cleavage of proteins for amino acid sequence determination or peptide mapping

  • Unit Definition: One unit will hydrolyze casein to change A280 0.001 per minute, pH 7.8, at 37°C.
  • Extinction Coefficient: (E1%) E1%280 = 4.26
  • Solubility: Soluble in aqueous buffers (such as 10 mM Tris, pH 7.8 at 1 mg/mL).
  • CAS Number: 66676-43-5
  • Molecular Weight: 27,700 (sedimentation equilibrium)
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Used for selective cleavage of Proteins for Amino Acid sequence determination or Peptide mapping
pH Range 4 and 7.8
Sample Type Staphylococcus aureus V8 Strain
Content And Storage -20°C
Form Powder
Quantity 5 mg
Product Type Protease
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.